Learn More
Kingfisher Biotech Inc Chicken IL-6 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0299C005
Encompass
The Chicken IL-6 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken IL-6 applications are for cell culture, ELISA standard, and Western Blot Control. The Chicken IL-6 yeast-derived recombinant protein can be purchased in multiple sizes. Chicken IL-6 Specifications: (Molecular Weight: 22.0 kDa) (Amino Acid Sequence: VPLPAAADSS GEVGLEEEAG ARRALLDCEP LARVLRDRAV QLQDEMCKKF TVCENSMEML VRNNLNLPKV TEEDGCLLAG FDEEKCLTKL SSGLFAFQTY LEFIQETFDS EKQNVESLCY STKHLAATIR QMVINPDEVV IPDSAAQKSL LANLKSDKDW IEKITMHLIL RDFTSFMEKT VRAVRYLKKT RSFSA (195)) (Gene ID: 395337). For research use only. Made in the USA
Catalog No. 50-241-3833