Learn More
Raybiotech Inc Chicken IL-6 ELISA Kit
Supplier:  Raybiotech Inc ELGIL61
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.085 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Chicken IL-6 .
Catalog No. 50-194-8932