Learn More
American Research Products Inc Chicken Immunoglobulin of Yolk
Catalog No. 5000326366
Encompass
Chicken Immunoglobulin of Yolk IgY ELISA Kit from CUSABIO Detection Range 0 078 ng/mL-5 ng/mL Sensitivity 0 02 ng/mL Sample serum plasma egg yolk Method Sandwich
Catalog No. 50-003-26366 Supplier American Research Products Inc Supplier No. CSBE11635CH
Chicken Immunoglobulin of Yolk IgY ELISA Kit from CUSABIO Detection Range 0 078 ng/mL-5 ng/mL Sensitivity 0 02 ng/mL Sample serum plasma egg yolk Method Sandwich