Learn More
American Research Products Inc Chicken Insulin like growth fa
Catalog No. 5000328907
Encompass
Chicken Insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 125 pg/mL-2000 pg/mL Sensitivity 125 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28907 Supplier American Research Products Inc Supplier No. CSBE09867CH
Chicken Insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 125 pg/mL-2000 pg/mL Sensitivity 125 pg/mL Sample serum plasma tissue homogenates Method Competitive