Learn More
Biomatik Corporation Chicken Interferon Alpha (IFNa) CLIA Kit
Supplier:  Biomatik Corporation EKU09636
Encompass_Preferred
Chicken Interferon Alpha (IFNa) CLIA Kit for detecting IFNa in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.52pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P42165. Gene ID: 100857744. Target Synonyms: IFNA1; IFL; LeIF D; IFN; IFN-Alpha; IFNA13; IFN-A; IFNAP22; IFN-Alpha 1b; Interferon Alpha 1b; Interferon, Leukocytic; IFN, Leukocyte; Interferon alpha-D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5533