Learn More
Biomatik Corporation Chicken Interferon Beta (IFNb) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10287
Encompass_Preferred
Chicken Interferon Beta (IFNb) High-sensitive ELISA Kit for detecting IFNb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.16pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q90873. Gene ID: 554219. Target Synonyms: IFNB1; IFN-B; IFB; IFF; IFNB; Interferon Beta 1 Fibroblast. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8352