Learn More
American Research Products Inc Chicken Interleukin 10 IL 10
Catalog No. 5000329949
Encompass
Chicken Interleukin 10 IL-10 ELISA Kit from CUSABIO Detection Range 1 pg/mL-200 pg/mL Sensitivity 0 5 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29949 Supplier American Research Products Inc Supplier No. CSBE12835C
Add to Cart
Add to Cart
Chicken Interleukin 10 IL-10 ELISA Kit from CUSABIO Detection Range 1 pg/mL-200 pg/mL Sensitivity 0 5 pg/mL Sample serum plasma tissue homogenates Method Competitive