Learn More
American Research Products Inc Chicken Interleukin 12 IL 12
Supplier:  American Research Products Inc CSBE12836C
Encompass
Chicken Interleukin 12 IL-12/70 ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26249