Learn More
American Research Products Inc Chicken Interleukin 17 IL 17
Catalog No. 5000326377
Encompass
Chicken Interleukin 17 IL-17 ELISA Kit from CUSABIO Detection Range 150 pg/mL-6000 pg/mL Sensitivity 150 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26377 Supplier American Research Products Inc Supplier No. CSBE04607CH
Chicken Interleukin 17 IL-17 ELISA Kit from CUSABIO Detection Range 150 pg/mL-6000 pg/mL Sensitivity 150 pg/mL Sample serum plasma tissue homogenates Method Competitive