Learn More
American Research Products Inc Chicken Interleukin 8 IL 8 E
Supplier:  American Research Products Inc CSBE14191C
Encompass
Chicken Interleukin 8 (IL-8) ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-400 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29577