Learn More
Kingfisher Biotech Inc Chicken LIF Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1395C025
Encompass
The Chicken Leukemia Inhibitory Factor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken Leukemia Inhibitory Factor applications are for cell culture, ELISA standard, and Western Blot Control. Chicken Leukemia Inhibitory Factor yeast-derived recombinant protein can be purchased in multiple sizes. Chicken Leukemia Inhibitory Factor Specifications: (Molecular Weight: 21.0 kDa) (Amino Acid Sequence: RALLGTSSAC PTNGLCRANV LEQTRRQVAL LNATAQDLFS LYLKCQGEPF SSESDRLCSP SGIFFPPFHV NRTTERKEVM VAMYKLFAFL NASLGNITRD QEELNPMAKE LLNRLHNTTK TTRGLISNLT CLLCKHYNIF QVDVSYGESS KDKSAFKKKQ QGCQVLRKYV QVIAQAARVL LPHLSPA (187)) (Gene ID: 427718). For research use only. Made in the USA
Catalog No. 50-253-3312