Learn More
Biomatik Corporation Chicken Lysozyme (Lzm) Elisa Kit
Supplier:  Biomatik Corporation EKC31041
Encompass_Preferred
Chicken Lysozyme (Lzm) Elisa Kit for detecting Lysozyme (Lzm) in serum, plasma, egg white. Species Reactivity: Chicken (Gallus). Detection Range: 15.625-4000ng/ml. Sensitivity: 14.876ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Target Synonyms: LZM; lysozyme; 1; 4-beta-N-acetylmuramidase C; lysozyme; lysozyme C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9199