Learn More
Biomatik Corporation Chicken Lysozyme (LZM) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11216
Encompass_Preferred
Chicken Lysozyme (LZM) Mini sample ELISA Kit for detecting LZM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.293ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00698. Gene ID: 396218. Target Synonyms: LYZ; Renal Amyloidosis; N-Acetylmuramide Glycanhydrolase; Muramidase; 1,4-beta-N-acetylmuramidase C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8195