Learn More
Kingfisher Biotech Inc Chicken M-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1335C100
Encompass
The Chicken M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Chicken M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Chicken M-CSF Specifications: (Molecular Weight: 27.2 kDa) (Amino Acid Sequence: NSYCQQIITE RHLDHLQELA DTQMQQPGTV SFRFISKMRL SDSVCYVKAA FPLLGTILNR TTFKENSTNA NKMKTVRKMY ENIDEDVDPC IRDEDDEEHA LSEMCFEEFT TSPYEMLVLV RQFFQDIKQL LQNKETFEKD CSQVYRSACA GPRQHSSSPE QRRKEPPAAS GGHRELLVYV TVASVVAVLL AMGGLLFYKY KSKVLQRGAA LKEGGCDPEE PESRALQGAQ GCAELETQEL (240)) (Gene ID: 100499189). For research use only. Made in the USA
Catalog No. 50-253-3359