Learn More
Kingfisher Biotech Inc Chicken M-CSF Polyclonal Antibody - Biotinylated (50 ug)
SDP
Supplier:  Kingfisher Biotech Inc KPB1522C050
Encompass
The Chicken M-CSF polyclonal antibody is labeled and has been qualified for use in ELISA. It is the detection antibody in the Kingfisher Chicken M-CSF ELISA. The Chicken M-CSF antibody was produced in rabbits and is antigen-affinity purified. The reactivity by species is: (Bovine M-CSF - None) (Chicken M-CSF - Strong) (Mouse M-CSF - None). For research use only. Made in the USA
Catalog No. 50-233-2553