Learn More
Biomatik Corporation Chicken Matrix Metalloproteinase 13 (MMP13) ELISA Kit
Supplier:  Biomatik Corporation EKU05835
Encompass_Preferred
Chicken Matrix Metalloproteinase 13 (MMP13) ELISA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 1.56-100ng/ml. Sensitivity: 5.7ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1NZ56. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8881