Learn More
Biomatik Corporation Chicken Matrix Metalloproteinase 13 (MMP13) CLIA Kit
Supplier:  Biomatik Corporation EKU09573
Encompass_Preferred
Chicken Matrix Metalloproteinase 13 (MMP13) CLIA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: F1NZ56. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8494