Learn More
Biomatik Corporation Chicken Myostatin (MSTN) CLIA Kit
Supplier:  Biomatik Corporation EKU09683
Encompass_Preferred
Chicken Myostatin (MSTN) CLIA Kit for detecting MSTN in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O42220. Gene ID: 373964. Target Synonyms: GDF8; Growth Differentiation Factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8467