Learn More
Biomatik Corporation Chicken Nuclear Factor Kappa B (NFkB) ELISA Kit
Supplier:  Biomatik Corporation EKU06351
Encompass_Preferred
Chicken Nuclear Factor Kappa B (NFkB) ELISA Kit for detecting NFkB in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04861. Gene ID: 396033. Target Synonyms: NF-KB1; EBP-1; KBF1; NF-Kappa-B; NFKB-P105; NFKB-P50; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1; Nuclear factor NF-kappa-B p105 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8829