Learn More
Encor Biotechnology Inc Chicken Polyclonal Antibody to GFP, 100 uL
Supplier:  Encor Biotechnology Inc CPCAGFP 100UL
Encompass
The CPCA-GFP antibody was made against a recombinant GFP construct originating from an Aequoria species which was engineered to improve spectral properties and prevent oligomerization. This form of GFP, referred to as AcGFP, is 94% identical to the eGFP developed by Tsien and coworkers and is the form of GFP inserted in the Clontech/Takara pAcGFP and related expression vectors.
Catalog No. NC1798346