Learn More
American Research Products Inc Chicken progesterone PROG ELI
Catalog No. 5000330032
Encompass
Chicken progesterone PROG ELISA Kit from CUSABIO Detection Range 1 ng/ml-70 ng/ml Sensitivity 0 2 ng/ml Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-30032 Supplier American Research Products Inc Supplier No. CSBE12012C
Chicken progesterone PROG ELISA Kit from CUSABIO Detection Range 1 ng/ml-70 ng/ml Sensitivity 0 2 ng/ml Sample serum plasma tissue homogenates Method Competitive