Learn More
Innovative Research Inc Chicken Red Blood Cells Packed 5% 30ml
Catalog No. 502034863
Encompass_Preferred
Chicken Red Blood Cells from Innovative Research are manufactured using whole blood collections from chickens. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 5% solution in Alsever's. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4863 Supplier Innovative Research Inc Supplier No. ICHRBC5P30ML
Chicken Red Blood Cells from Innovative Research are manufactured using whole blood collections from chickens. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 5% solution in Alsever's. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.