Learn More
Innovative Research Inc Chicken Red Blood Cells Packed 5% 50ml
Supplier:  Innovative Research Inc ICHRBC5P50ML
Encompass_Preferred
Chicken Red Blood Cells from Innovative Research are manufactured using whole blood collections from chickens. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 5% solution in Alsever's. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4869
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.