Learn More
Biomatik Corporation Chicken Retinol Binding Protein 4 (RBP4) ELISA Kit
Supplier:  Biomatik Corporation EKU11402
Encompass_Preferred
Chicken Retinol Binding Protein 4 (RBP4) ELISA Kit for detecting RBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.071ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41263. Gene ID: 396166. Target Synonyms: PRBP; RBP; Plasma retinol-binding protein; Retinol Binding Protein 4, Plasma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5222