Learn More
American Research Products Inc Chicken rudimental boVine seru
Catalog No. 5000327369
Encompass
Chicken rudimental boVine serum albumin (BSA) check-up ELISA kit from CUSABIO Detection Range 0 312 g/mL-80 g/mL Sensitivity 0 312 g/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27369 Supplier American Research Products Inc Supplier No. CSBEQ027303CH
Chicken rudimental boVine serum albumin (BSA) check-up ELISA kit from CUSABIO Detection Range 0 312 g/mL-80 g/mL Sensitivity 0 312 g/mL Sample serum plasma tissue homogenates Method Competitive