Learn More
Kingfisher Biotech Inc Chicken SCF Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1555C005
Encompass
The Chicken SCF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Chicken SCF applications are for cell culture, ELISA standard, and Western Blot Control. Chicken SCF yeast-derived recombinant protein can be purchased in multiple sizes. Chicken SCF Specifications: (Molecular Weight: 22.5 kDa) (Amino Acid Sequence: QSSCGNPVTD DVNDIAKLVG NLPNDYLITL KYVPKMDSLP NHCWLHLMVP EFSRSLHNLL QKFSDISDMS DVLSNYSIIN NLTRIINDLM ACLAFDKNKD FIKENGHLYE EDRFIPENFF RLFNSTIEVY KEFADSLDKN DCIMPSTVET PENDSRVAVT KTISFPPVAA SSLRNDSIGS NTSSNSNKEA LGFISSSSLQ (200)) (Gene ID: 396028). For research use only. Made in the USA
Catalog No. 50-253-3149