Learn More
American Research Products Inc Chicken secretory immunoglobul
Catalog No. 5000326062
Encompass
Chicken secretory immunoglobulin A SIgA ELISA Kit from CUSABIO Detection Range 0 234 g/mL-60 g/mL Sensitivity 0 058 g/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26062 Supplier American Research Products Inc Supplier No. CSBE10097CH
Add to Cart
Add to Cart
Chicken secretory immunoglobulin A SIgA ELISA Kit from CUSABIO Detection Range 0 234 g/mL-60 g/mL Sensitivity 0 058 g/mL Sample serum plasma Method Competitive