Learn More
Equitech Bio Inc Chicken Serum Albumin, Lyophilized Powder. 10mg.
SDP
Catalog No. NC3312202
Encompass
Chicken Serum Albumin, Lyophilized Powder. 10mg. Prepared from chicken serum by heat-shock fractionation. Country of origin: USA. Purity: ≥ 96%
Catalog No. NC3312202 Supplier Equitech Bio Inc Supplier No. CSA620010
Chicken Serum Albumin, Lyophilized Powder. 10mg. Prepared from chicken serum by heat-shock fractionation. Country of origin: USA. Purity: ≥ 96%