Learn More
Biomatik Corporation Chicken Serum Amyloid A (SAA) ELISA Kit
Supplier:  Biomatik Corporation EKU11424
Encompass_Preferred
Chicken Serum Amyloid A (SAA) ELISA Kit for detecting SAA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1NW65. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4880