Learn More
Biomatik Corporation Chicken Superoxide Dismutase 1 (SOD1) ELISA Kit
Supplier:  Biomatik Corporation EKU12189
Encompass_Preferred
Chicken Superoxide Dismutase 1 (SOD1) ELISA Kit for detecting SOD1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80566. Gene ID: 395938. Target Synonyms: ALS; ALS1; IPOA; Superoxide Dismutase 1, Soluble; Amyotrophic Lateral Sclerosis 1,Adult; Superoxide dismutase [Cu-Zn]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8012