Learn More
Kingfisher Biotech Inc Chicken M-CSF ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891890
Encompass_Preferred
Chicken M-CSF ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1890 Supplier Kingfisher Biotech Inc Supplier No. SPOT2408C004
Chicken M-CSF ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA