Learn More
American Research Products Inc Chicken Transforming Growth fa
Catalog No. 5000328969
Encompass
Chicken Transforming Growth factor b1 TGF-b1 ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28969 Supplier American Research Products Inc Supplier No. CSBE09875CH
Chicken Transforming Growth factor b1 TGF-b1 ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich