Learn More
American Research Products Inc Chicken Tri iodothyronine T3
Catalog No. 5000329827
Encompass
Chicken Tri-iodothyronine T3 ELISA Kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29827 Supplier American Research Products Inc Supplier No. CSBE13270C
Chicken Tri-iodothyronine T3 ELISA Kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive