Learn More
Biomatik Corporation Chicken Trypsin (TRY) ELISA Kit
Supplier:  Biomatik Corporation EKU11494
Encompass_Preferred
Chicken Trypsin (TRY) ELISA Kit for detecting TRY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 78-5,000pg/ml. Sensitivity: 35pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q90627. Gene ID: 396345. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5607