Learn More
American Research Products Inc Chicken Tumor necrosis factor
Catalog No. 5000328997
Encompass
Chicken Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 0 27 pg/mL-200 pg/mL Sensitivity 0 27 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28997 Supplier American Research Products Inc Supplier No. CSBE11231CH
Chicken Tumor necrosis factor a TNF-a ELISA KIT from CUSABIO Detection Range 0 27 pg/mL-200 pg/mL Sensitivity 0 27 pg/mL Sample serum plasma tissue homogenates Method Competitive