Learn More
Bioss Chickungunya virus
Supplier: Bioss BSM72042M
Chickungunya virus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide