Learn More
Enterprise Technology Solutions Chicopee Branded Products Dust Cloth, StretchnDust,12"x17",40/PK,10PK/CT,YW/OE
SDP
Catalog No. 5000810422
Encompass_Preferred
Stretch 'N Dust Medium-Duty Cloths are ideal for multipurpose dusting. They are strong and durable yet soft enough for delicate surfaces. Low-linting cloths are made of viscose fibers and pre-impregnated with mineral oil to create a dust-catching, 3-D structure. They hold the smallest particles in the treated fabric.
Catalog No. 50-008-10422 Supplier Enterprise Technology Solutions Supplier No. CHI0413
Stretch 'N Dust Medium-Duty Cloths are ideal for multipurpose dusting. They are strong and durable yet soft enough for delicate surfaces. Low-linting cloths are made of viscose fibers and pre-impregnated with mineral oil to create a dust-catching, 3-D structure. They hold the smallest particles in the treated fabric.