Learn More
Enterprise Technology Solutions Chicopee Branded Products Towels,Shop,Heavy-Duty,Washable,Cloth,13.5"x15",300/CT,White
SDP
Catalog No. 5000810423
Encompass_Preferred
Heavy-duty shop towels will effectively replace your worn-out laundered shop towels used in heavy-duty, general-purpose applications. Thick, durable cloth quickly absorbs water and oil spills without tearing, even on rough surfaces. Easy-to-rinse, low-linting towels designed for multiple uses stand up to at least 50 machine-washes. Substantial draping lets you remove shop towels from packaging with ease.
Catalog No. 50-008-10423 Supplier Enterprise Technology Solutions Supplier No. CHI8483
Heavy-duty shop towels will effectively replace your worn-out laundered shop towels used in heavy-duty, general-purpose applications. Thick, durable cloth quickly absorbs water and oil spills without tearing, even on rough surfaces. Easy-to-rinse, low-linting towels designed for multiple uses stand up to at least 50 machine-washes. Substantial draping lets you remove shop towels from packaging with ease.