Learn More
Cayman Chemical ChIcorIc AcId 10mg
SDP
Catalog No. 502934129
Encompass_Preferred
A dicaffeoyl ester with diverse biological activities; reduces the viability of Caco-2 and HCT116 colorectal cancer cells from 50-200 μg/ml; inhibits the 3'-processing and DNA integration activities of HIV integrase (IC50s = 1.1 and 0.8 μM, respectively); inhibits HIV entry into H9 cells at 5 μM; oral administration reduces hepatic lipid accumulation, lipid peroxidation, and fibrosis, inhibits production of pro-inflammatory cytokines and activation of NF-kB, and activates the AMPK signaling pathway in a mouse model of NASH induced by a methionine and choline-deficient diet at 10 and 30 mg/kg; reduces blood glucose levels by 54% in mice with streptozotocin-induced diabetes at 2 mg/kg
Catalog No. 50-293-4129 Supplier Cayman Chemical Supplier No. 2496010 MG
A dicaffeoyl ester with diverse biological activities; reduces the viability of Caco-2 and HCT116 colorectal cancer cells from 50-200 μg/ml; inhibits the 3'-processing and DNA integration activities of HIV integrase (IC50s = 1.1 and 0.8 μM, respectively); inhibits HIV entry into H9 cells at 5 μM; oral administration reduces hepatic lipid accumulation, lipid peroxidation, and fibrosis, inhibits production of pro-inflammatory cytokines and activation of NF-kB, and activates the AMPK signaling pathway in a mouse model of NASH induced by a methionine and choline-deficient diet at 10 and 30 mg/kg; reduces blood glucose levels by 54% in mice with streptozotocin-induced diabetes at 2 mg/kg