Learn More
Ted Pella Inc Chien Staining Pad, White, each
SDP
Catalog No. NC1407537
Encompass
The Chien Staining Pad is also ideal for staining grids in the microwave processor. The silicone pad is flexed and tabbed electron microscopy grids are inserted into the slits. When the pad is unflexed (laid flat) the grids stand vertically, allowing the surface tension of a droplet of stain to cover both sides of grid.
Catalog No. NC1407537 Supplier Ted Pella Inc Supplier No. 105232
The Chien Staining Pad is also ideal for staining grids in the microwave processor. The silicone pad is flexed and tabbed electron microscopy grids are inserted into the slits. When the pad is unflexed (laid flat) the grids stand vertically, allowing the surface tension of a droplet of stain to cover both sides of grid.