Learn More
Electron Microscopy Sciences Chien Universal-Mold
SDP
Catalog No. 5024962
Encompass_Preferred
A two-directional mold that allows for single embedded specimens to be sectioned both laterally and transversely . Tissues to be embedded are accurately trimmed and placed so that both sides of the tissue are against both edges of the cavity tip. See Chien, K., et al. (1985) Proc. Ann. EMSA Meeting 43, 460
Catalog No. 50-249-62 Supplier Electron Microscopy Sciences Supplier No. 70141
A two-directional mold that allows for single embedded specimens to be sectioned both laterally and transversely . Tissues to be embedded are accurately trimmed and placed so that both sides of the tissue are against both edges of the cavity tip. See Chien, K., et al. (1985) Proc. Ann. EMSA Meeting 43, 460