Learn More
Research Products International Corp Lab Armor Chill Bucket with Beads and 2 Chill Packs, 1 Package
SDP
Catalog No. 50550335
Encompass
This innovative chill bucket, with lid and bead bag, keeps samples at -20°C to +8°C for up to eight hours while on a benchtop. Supplied with 2 liters thermal beads and chill packs that eliminate ice and maintain a dry and clean environment. Beads also keep tubes and vessels in place without use of racks.
Catalog No. 50-550-335 Supplier Research Products International Corp Supplier No. 299701
This innovative chill bucket, with lid and bead bag, keeps samples at -20°C to +8°C for up to eight hours while on a benchtop. Supplied with 2 liters thermal beads and chill packs that eliminate ice and maintain a dry and clean environment. Beads also keep tubes and vessels in place without use of racks.