Learn More
Ergodyne™ Chill-Its™ 5155 Low Profile Hydration Pack
Catalog No. 19819776 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Hi-vis Orange
Hi-vis Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-776 Black
19-819-777 Hi-vis Yellow
19-819-778 Hi-vis Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-819-776 Supplier Ergodyne™ Supplier No. 13155
Add to Cart
Add to Cart
Low Profile Hydration Pack helps to keep workers hydrated when they're away from a water source.
  • Durable 600D water-resistant rip stop polyester shell
  • Zip-up cap cover keeps debris and contaminants at bay
  • Insulated pack and tube keeps water cool
  • 100% anti-microbial bite valve with cover to protect against contaminants
  • Bite valve cover for protection from contaminants
  • Breakaway shoulder straps for added safety
  • DualCap bladder featuring smaller (60 mm) cap nestled into larger (80 mm) cap for easy filling
Specifications
Material 600D Polyester
Color Black
Closure Type DualCap
Capacity 2 L