Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6215 Premium FR Phase Change Cooling Vest with Packs

Catalog No. 19910201 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Orange
Tan
Size:
Large/X-Large
Small/Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-910-201 Orange Large/X-Large
19-819-668 Tan Small/Medium
19-819-687 Tan Large/X-Large
19-819-688 Orange Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-910-201 Supplier Ergodyne™ Supplier No. 12221
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chill-Its™ 6215 Premium Phase Change Cooling Vest with Packs keeps workers cool and safe while maintaining a cool temperature for up to 4 hours when activated.

  • Immediate and long lasting cooling relief
  • Packs maintain constant 64°F (18°C) for up to 4 hours
  • Quick recharge in a little as 10 minutes
  • Honeycomb design eliminates stiffness
  • Flame resistant
  • Vest made with modacrylic cotton blend FR fabric
  • Flexible fit
  • Adjustable hook and loop closures on both shoulders and waist
  • Extended vest surface area enhances cooling power
  • Designed for more visibility on busy streets or job sites
  • Features two elastic pen loops on chest

Specifications

Material Modacrylic Cotton Blend FR Fabric
Product Type Cooling Vest
Size Large/X-Large
Length (Metric) 102 to 132 cm
Length (English) 40 to 52 in.
Color Orange
Certifications/Compliance ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); Classified to NFPA 70E (CAT1)
Cooling Time Packs Maintain Constant 64°F (18°C) for up to 4 hours
For Use With (Application) Fire Rescue, EMT, Construction, Utilities
Includes Cooling Packs, Adjustable Hooks, and Loop Closures at Shoulders
Quantity 1/Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.