Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6220 Phase Change Cooling Vest Charge Packs

Catalog No. 19819672 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-672 Large/X-Large
19-819-671 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-672 Supplier Ergodyne™ Supplier No. 12202
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chill-Its™ 6220 Phase Change Cooling Vest Charge Packs are suitable for use with a variety of cooling vests.

  • Immediate and long-lasting cooling relief
  • Packs maintain constant 64°F (18°C) for up to 4 hours
  • Quick recharge in a little as 10 minutes
  • Patented honeycomb ice pack design eliminates stiffness
  • Wash by hand with a mild detergent and warm water

Specifications

Material Cotton with Gel Polymer
Product Type Cooling Pack
Size Large/X-Large
Color Clear
Cooling Time Packs Maintain Constant 64°F (18°C) for up to 4 hours
For Use With (Application) Cooling Vest
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.