Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6605 High-Performance Headband

Catalog No. 19819513 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Camouflage
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-513 Blue
19-819-509 Camouflage
19-819-516 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-819-513 Supplier Ergodyne™ Supplier No. 12425
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-Performance Headband constructed of technical fabric to absorb moisture away from the body for fast evaporation.

  • Hi Cool™ moisture management performance fabric
  • Absorbent terry headband
  • Elasticized back for a great fit
  • Available in the following patterns: Stars and Stripes, Camouflage, Red Western, Navy Western, Blue, Lime, Flames, and Skulls

Specifications

Material Hi Cool™ Moisture Management Performance Fabric
Color Blue
For Use With (Application) Construction, Maintenance, Landscaping
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.