Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6630 High-Performance Cap

Catalog No. 19819538 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Blue
Blue, White
Gray, Black, White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-538 Blue
19-819-539 Black
19-819-545 Gray, Black, White
19-819-536 Blue, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-819-538 Supplier Ergodyne™ Supplier No. 12510
Add to Cart
Add to Cart

High-Performance Cap constructed of technical fabric with a wide, absorbent terry headband is comfortable to wear under a helmet or alone.

  • Hi Cool™ moisture management performance fabric
  • Comfortable to wear under hats, helmets or alone
  • Wide, absorbent terrycloth band
  • One size fits most
  • Available in a varisty of colors and patterns like blue, black, skulls, and navy western

Specifications

Material Hi Cool™ Moisture Management Performance Fabric
Size Universal
Color Blue
For Use With (Application) Construction, Maintenance, Landscaping, Distribution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.