Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6630 High-Performance Cap

Catalog No. 19154630 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Camouflage
Flames
Hi-vis Lime
Red Western
Stars and Stripes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-154-630 Camouflage
19-154-628 Hi-vis Lime
19-154-629 Stars and Stripes
19-154-631 Red Western
19-154-635 Flames
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-154-630 Supplier Ergodyne™ Supplier No. 12507
Add to Cart
Add to Cart
This item is not returnable. View return policy

Comfortable alone or under helmet

  • Hi Cool™ Moisture Management performance fabric
  • Comfortable to wear under hats, helmets, or alone
  • Wide, absorbent terrycloth band
  • Elastic band for a great fit

Specifications

Material Fabric
Product Type High Performance Cap
Color Camouflage
For Use With (Application) Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, hells kitchen
For Use With (Equipment) For use under hats, helmets, or alone
Includes Elastic Band
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.