Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6650 High Performance Hat with Neck Shade

Catalog No. 19819543 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Lime
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-543 Orange
19-819-542 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-543 Supplier Ergodyne™ Supplier No. 12521
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-Performance Hat with Neck Shade is a machine washable hat designed to get moisture away from skin.

  • Hi Cool™ moisture management performance fabric
  • Absorbent terry headband
  • Wide panel neck protection
  • Adjustable cinch cord for a great fit

Specifications

Material Hi Cool™ Moisture Management Performance Fabric
Product Type Headband
Color Orange
Closure Type Adjustable Cinch Cord
For Use With (Application) Construction, Maintenance, Landscaping, Distribution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.