Learn More
Ergodyne™ Chill-Its™ 6685 Premium Dry Evaporative Cooling Vest
Catalog No. 19910210 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
6/Pk.
Catalog No. Quantity
19-910-210 6/Pk.
1 options
Catalog No. 19-910-210 Supplier Ergodyne™ Supplier No. 12678
Chill-Its™ 6685 Premium Dry Evaporative Cooling Vest offers immediate dry cooling relief that lasts up to three days.
  • Dry technology keeps wearer completely dry and comfortable during the entire evaporative cooling process
  • Up to 3 days of cooling power to help maintain a healthy body temperature
  • Fill vest with 13 to 20 oz. (400 to 600 mL) of water
  • Made of industrial grade quality nylon
  • Mesh side-panels provide additional ventilation and stretch
  • V-neck design with front zipper closure
  • Cooling material is treated with an anti-microbial that prevents mold growth
  • Reduces the effect of heat exhaustion and fights fatigue while working in the outdoor heat
  • Machine washable
Specifications
Size 4X-Large
Color Lime
For Use With (Application) Construction, Trades, Maintenance, Landscaping, Material Handling,Freight/Baggage, Warehousing/Distribution, Iron/Steel Fabrication
Quantity 6/Pk.